Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
10 ea | Warenkorb auf Verfügbarkeit prüfen | € 182,00 |
100 ea | Warenkorb auf Verfügbarkeit prüfen | € 1.760,00 |
Über diesen Artikel
€ 182,00
material
polysulfone , round bottom tube, screw top cap (Linerless), transparent tube
feature
RCF 50,000 × g
packaging
pack of 10, case of 100
manufacturer/tradename
Nalgene 3115-0050
nominal capacity
50 mL
General description
Features and Benefits
- Round bottoms to fit high speed rotors.
- Transparent with straw-colored cast.
- Comes with polypropylene linerless closures—easy to clean and provides leakproof† protection under normal conditions up to 10,000 x g.
- When equipped with a Nalgene sealing cap (sold separately), 30mL and 50mL tubes can be spun up to 50,000 x g.
- Can be used in refrigerated or non-refrigerated centrifuges.
- Tubes must be filled to at least 80% capacity for proper performance.
- Autoclavable/Leakproof† when used with supplied Nalgene closure.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description nominal capacity 50 mL | description nominal capacity 30 mL | description nominal capacity 50 mL | description nominal capacity 50 mL |
| manufacturer/tradename Nalgene 3115-0050 | manufacturer/tradename Nalgene 3115-0030 | manufacturer/tradename Nalgene® 3118-0050 | manufacturer/tradename Nalgene® 3119-0050 |
| material screw top cap (Linerless), round bottom tube, transparent tube, polysulfone | material transparent tube, screw top cap (Linerless), round bottom tube, polysulfone | material transparent polycarbonate , polypropylene screw closure, round bottom tube, screw top cap | material polypropylene screw closure, polypropylene copolymer , screw top cap (Linerless), round bottom tube |
| feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 100,605 × g |
| packaging pack of 10, case of 100 | packaging case of 100, pack of 10 | packaging pack of 10 | packaging pack of 10 ea |
| nominal capacity 50 mL | nominal capacity 30 mL | nominal capacity 50 mL | nominal capacity 50 mL |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge tubes, Oak Ridge style 3115
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?