Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 1.130,00 |
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121500
Volume:
10 μL
Packaging:
pack of 1 ea
Feature:
8-channel, electronic: no
Manufacturer/tradename:
Sartorius 725120
€ 1.130,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenfeature
8-channel, electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 725120
volume measuring range
0.5-10 μL
volume
10 μL
General description
Sartorius′s new mechanical pipettor family - the mLINE - offers a unique combination of novel features for mechanical pipetting. Excellent performance, full autoclavability, smooth ergonomics and perfect safety. The mLINE pipettors cover the full volume range of 0.1 to 5000.
- Extremely light pipetting and tip ejection
- Unique safety features
- Stepless volume adjustment and patented volume locking system
- Thermal insulation of internal components to improve accuracy
- Full autoclavability without disassembling
- Easy cleaning, maintenance and calibration
- ID tag and color-coded caps
- Safe-Cone Filters and unique ejection
- Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 8 channel, volume 0.5-10 μL | description 12 channel, volume 5-100 μL | description 12 channel, volume 0.5-10 μL | description 8 channel, volume 5-100 μL |
| manufacturer/tradename Sartorius 725120 | manufacturer/tradename Sartorius 725230 | manufacturer/tradename Sartorius 725220 | manufacturer/tradename Sartorius 725130 |
| volume 10 μL | volume 100 μL | volume 10 μL | volume 100 μL |
| feature 8-channel, electronic: no | feature electronic: no, 12-channel | feature electronic: no, 12-channel | feature 8-channel, electronic: no |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| volume measuring range 0.5-10 μL | volume measuring range 5-100 μL | volume measuring range 0.5-10 μL | volume measuring range 5-100 μL |
Still not finding the right product?
Explore all of our products under Sartorius mLINE® mechanical pipettor
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?