Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
10 ea | Warenkorb auf Verfügbarkeit prüfen | € 332,00 |
Über diesen Artikel
€ 332,00
material
(polycaprolactone (PCL))
sterility
sterile
feature
~ 300 μm spacing
manufacturer/tradename
3D Biotek PCL303006-BR
fiber diam.
~300 μm
application(s)
3D cell culture
binding type
non-treated surface
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description fiber diam. ~300 μm, ~ 300 μm spacing | description for 3D bioreactor 24 well chamber, fiber diam. ~300 μm | description for 3D bioreactor 12 well chamber, fiber diam. ~300 μm, ~ 300 μm spacing | description for 96 well plates, 24 inserts/ 96 well plate |
| binding type non-treated surface | binding type non-treated surface | binding type non-treated surface | binding type non-treated surface |
| manufacturer/tradename 3D Biotek PCL303006-BR | manufacturer/tradename 3D Biotek PCL303024-BR | manufacturer/tradename 3D Biotek PCL303012-BR | manufacturer/tradename 3D Biotek PCL303096-24 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material (polycaprolactone (PCL)) | material - | material (polycaprolactone (PCL)) | material clear polycaprolactone (PCL) , clear wells |
| feature ~ 300 μm spacing | feature - | feature ~ 300 μm spacing, lid: no | feature - |
| fiber diam. ~300 μm | fiber diam. ~300 μm | fiber diam. ~300 μm | fiber diam. - |
Still not finding the right product?
Explore all of our products under 3D Biotek Polycaprolactone (PCL) scaffold inserts
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?