Fortfahren mit
Z694495
Nanofiber cell culture dish, with random oriented nanofibers
diam. 100 mm
Synonym(e):
3d plates, tissue culture plates, Nanofiber Solutions 10001
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 83,40
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121812
Material:
round
Sterility:
sterile
Diam.:
100 mm
Binding type:
(plasma surface treated)
Manufacturer/tradename:
Nanofiber Solutions 10001
Packaging:
pack of 1 ea
€ 83,40
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
round
sterility
sterile
feature
lid: no
packaging
pack of 1 ea
manufacturer/tradename
Nanofiber Solutions 10001
diam.
100 mm
suitability
suitable for (Tissue Engineering, Cancer Research)
binding type
(plasma surface treated)
General description
Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for Cancer Research. The Cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z694487Z694592Z759341
description
diam. 100 mm
description
diam. 60 mm
description
diam. 100 mm
description
size 6 wells
binding type
(plasma surface treated)
binding type
(plasma surface treated)
binding type
(plasma surface treated)
binding type
non-treated surface
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
manufacturer/tradename
Nanofiber Solutions 10001
manufacturer/tradename
Nanofiber Solutions 6001
manufacturer/tradename
Nanofiber Solutions 10002
manufacturer/tradename
Nanofiber Solutions 601
feature
lid: no
feature
lid: no
feature
lid: no
feature
lid
material
round
material
round
material
round
material
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
-
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)