Fortfahren mit
Z688797
Nunc® UpCell Surface cell culture multidish
size 12 wells, case of 6 ea
Synonym(e):
cell culture multidish, tissue culture multidish, Nunc 174900, cultivation plate, surface treated dish
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
6 ea
Warenkorb auf Verfügbarkeit prüfen
€ 377,00
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121812
Material:
clear plate, clear wells
Sterility:
sterile
Binding type:
(UpCell Surface)
Manufacturer/tradename:
Nunc 174900
Packaging:
case of 6 ea
€ 377,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
clear plate, clear wells
sterility
sterile
feature
membrane not included, w/ lid and airvent, lid
packaging
case of 6 ea
manufacturer/tradename
Nunc 174900
dish L × W
128 mm × 86 mm
external L × W
128 mm × 86 mm
size
12 wells
surface area
3.5 cm2 , culture area (cm2)/well
working volume
2 mL
binding type
(UpCell Surface)
General description
The Nunc UpCell Surface dishes have a temperature responsive cell culture surface. These UpCell Surface dishes are suitable for harvesting of cells with intact surface proteins, for culture passaging, single-cell analyses and cell transplantation research. The UpCell surface enables harvesting of cell sheets and creation of 3D tissue models which are held together by normal cell junctions and extracellular matrix
Application
  • Culture passaging
  • Subsequent single-cell analyses (e.g. flow cytometry)
  • 3D tissue modeling for transplantation
Features and Benefits
  • The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
  • No trypsinization - preserve cell surface proteins
  • No need for dissociation enzymes, ensuring intact cell surface proteins
  • No need for physical scraping, resulting in high cell viability
  • Supportive membranes included in the 6-well plates allow for harvesting of the cell sheets
  • Enables harvesting of cell sheets and creations of 3D tissue models held together by normal cell junctions and extracellular matrix
  • Minimal hands-on time
  • For research and single-use only
  • It is quick, clean and simple-just reduce the temperature
  • Shelf Life: 3 years from date of manufacturing
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
UpCell is a trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z688800Z694347Z688827
description
size 12 wells, case of 6 ea
description
size 6 wells, case of 6 ea
description
size 96 wells, sterile, surface area 0.33 cm2
description
size 100 mm, w/ grid, sterile, case of 6 ea
feature
w/ lid and airvent, membrane not included
feature
lid, w/ membrane, w/ lid and airvent
feature
lid, membrane not included, w/ lid and airvent
feature
lid vented, w/ grid, membrane not included
binding type
(UpCell Surface)
binding type
(UpCell Surface)
binding type
(UpCell Surface)
binding type
(UpCell Surface)
manufacturer/tradename
Nunc 174900
manufacturer/tradename
Nunc 174901
manufacturer/tradename
Nunc 174897
manufacturer/tradename
Nunc 174905
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
clear wells, clear plate
material
clear plate, clear wells
material
clear polystyrene plate, rectangular clear wells
material
polystyrene
packaging
case of 6 ea
packaging
case of 6 ea
packaging
pack of 1 ea, case of 8 ea
packaging
case of 6 ea
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)