Fortfahren mit
Z688207
HandEvac handheld aspirator
8-channel adapter with permanent stainless steel tips, Cole Parmer 439607
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41105100
description
Adapter Only (hand operator not included)
manufacturer/tradename
Cole Parmer 439607
General description
Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes!
This ergonomic, hand-held aspirator can be used with pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques.
Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
8-Channel adapter has permanently fixed stainless steel tips.
Still not finding the right product?
Explore all of our products under HandEvac handheld aspirator
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)