Merck
Z659886
Bandelin SONOREX Ultrasonic bath accessories
positioning lid for Z659843 and Z659045
Größe auswählen
1 EA
€ 99,00
€ 99,00
Voraussichtliches Versanddatum30. Mai 2025
Größe auswählen
Ansicht ändern
1 EA
€ 99,00
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.74
€ 99,00
Voraussichtliches Versanddatum30. Mai 2025
Materialien
stainless steel
Leistungsmerkmale
for 4 beakers
Verpackung
pack of 1 ea
Hersteller/Markenname
Bandelin 382
Verfügbarkeit
not available in USA and Canada
Allgemeine Beschreibung
The insert strip is used to hold the insert containers. Insert cups are used when using chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids (indirect sonication). Because nothing can be placed on the bottom of the ultrasonic bath, suitable perforated lids hold the insert cups. When using an insert strip, several insert cups with different liquids can be sonicated at the same time. In order to transfer the ultrasound to the insert vessel, the surface tension of the water must be reduced and therefore a cleaning solution must be used as a coupling liquid / contact liquid.
Leistungsmerkmale und Vorteile
  • for holding insert cups
Rechtliche Hinweise
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)