Merck
Z645443
Nichiryo pipette tip loading system
volume range 0.1-10 μL, elongated, Reloading system, 10 x 96
About This Item
UNSPSC-Code:
41121600
Materialien
colorless
Sterilität
sterile
Leistungsmerkmale
barrier: no
Verpackung
pack of 960 ea
pack of 960
Hersteller/Markenname
Nichiryo BMT-SSEP
Verfügbarkeit
not available in (France)
Maximales Volumen
10 μL
Volumenbereich
0.1-10 μL
Allgemeine Beschreibung
Universal fit pipette tips supplied in racks with a convenient hinged lid giving quick easy access.
  • Manufactured RNAse and DNAse free
  • Eco-friendly design provides racked tips with minimum plastic waste
  • Can be used stacked or as individual racks
  • Convenient hinged rack provides quick, easy access to tips
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)