Fortfahren mit
Z617822
Greiner tissue culture tube with screw cap
capacity 14 mL
Synonym(e):
Greiner 163160, centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, sample receptacle, sample storage, sample tube, tube
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121800
Capacity:
14 mL
Material:
round bottom transparent polystyrene , screw top cap
Sterility:
sterile
Packaging:
pack of 1,000 ea
material
round bottom transparent polystyrene , screw top cap
sterility
sterile
packaging
pack of 1,000 ea
manufacturer/tradename
Greiner 163160
capacity
14 mL
diam. × L
16 mm × 100 mm
General description
Greiner CELLSTAR® is manufactured from crystal clearpolystyrene and has improved cell adhesion through physical surface treatment.Non-skirted, supplied with a red screw cap.
Features and Benefits
  • Sterile
  • Free detectable DNase, human DNA, RNase
  • Non cytotoxic and non pyrogenic
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)