Merck
Z380342
Slant rack for culture tubes
for 15-16 mm tubes
Synonym(e):
rack test tube, racks tube, test tube racks
Größe auswählen
1 EA
€ 93,80
4 EA
€ 278,00
€ 93,80
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 93,80
4 EA
€ 278,00
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.22
€ 93,80
Check Cart for Availability
Leistungsmerkmale
Holds 40 x 15-16 mm tubes
Verpackung
pack of 1 ea
case of 4 ea
Hersteller/Markenname
Nalgene® 5935-0016
Allgemeine Beschreibung
Unique end plate design allows 5° or 20° slant when rack is set down on side. Autoclave media upright, then allow to cool on a side. Media will harden with perfect, consistent slant. Also useful for incubating liquids on a slant for better aeration.
Rack holds 40 tubes (4 × 10 array); keyed with numbers and letters. Stackable; will not float in water bath. Autoclaving will reduce strength over time.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)