Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 29,10 |
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
glass
Packaging:
pkg of 1 ea
€ 29,10
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
glass
packaging
pkg of 1 ea
binding type
non-treated surface
General description
This is the replacement cover slip for the Bright-Line Hemacytometer, Prod. No. Z359629
This product consists of a single cover slip. The microscope slide cover slips are polished soda lime glass, they are rectangular in shape and measure: 20 mm x 26 mm x 0.4 mm thick.
This product consists of a single cover slip. The microscope slide cover slips are polished soda lime glass, they are rectangular in shape and measure: 20 mm x 26 mm x 0.4 mm thick.
Legal Information
Bright-Line is a trademark of Cambridge Instruments, Inc.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description replacement cover slip for the Bright-Line Hemacytometer: Z359629 | description supplied with two cover slips | description borosilicate glass, size 20 mm × 26 mm × 0.4 mm | description borosilicate glass, size 22 mm × 30 mm × 0.4 mm |
| material glass | material colorless , glass | material borosilicate glass | material borosilicate glass |
| packaging pkg of 1 ea | packaging - | packaging - | packaging - |
| binding type non-treated surface | binding type - | binding type non-treated surface | binding type non-treated surface |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?