Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 87,00 |
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121500
Manufacturer/tradename:
Bel-Art F37876-0000
Material:
polystyrene
€ 87,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polystyrene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37876-0000
L × W × diam.
127 mm × 86 × 37 mm
General description
A simple idea that saves a lot of time in a variety of multiwell protocols. For transfers to, from, and between 96 well plates, just connect the Vaccu-Pette/96 to a large syringe and aspirate about 100 times the desired per-well volume. For example, to transfer 50 μL/well, aspirate and deliver a total of 5 mL. Cells growing in the wells are protected by ridges that prevent access to the bottom 50 μl of each well. Ideal for ELISA, cytotoxicity, and cloning procedures. Without a syringe, the device will pick up about 15 μL per well, making it an effective 96 well blotting tool. 35 mL syringe not included.
Legal Information
Vaccu-Pette/96 is a trademark of Bel-Art Products, a division of SP
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description polystyrene | description non-sterile, polycarbonate, autoclavable | description vinyl bulb, fits 6-8 mm pipettes, L × diam. 70 mm × 5 mm | description 96 well plate for PCR |
| material polystyrene | material polycarbonate | material vinyl bulb | material colorless wells, colorless polypropylene |
| manufacturer/tradename Bel-Art F37876-0000 | manufacturer/tradename Bel-Art F37876-0001 | manufacturer/tradename Bel-Art F37887-0000 | manufacturer/tradename Sorenson 2633 |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 2 ea | packaging case of 25 ea |
| L × W × diam. 127 mm × 86 × 37 mm | L × W × diam. 127 mm × 86 mm × 37 mm | L × W × diam. - | L × W × diam. - |
Still not finding the right product?
Explore all of our products under Vaccu-Pette/96™ multiwell pipetter
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?