Fortfahren mit
Z332771
BRAND® Adapter für 25 & 50 ml PD-Tips
sterile
Synonym(e):
Adapter for Handystep tips
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 pkg
Warenkorb auf Verfügbarkeit prüfen
€ 23,80
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121600
Sterility:
sterile
Packaging:
pack of 5 ea
Manufacturer/tradename:
BRAND 702399
€ 23,80
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polypropylene
sterility
sterile
packaging
pack of 5 ea
manufacturer/tradename
BRAND 702399
maximum volume
50 mL
General description
The PD-Tips II dispenser tips for efficient, step-by-step dispensing enhance the proven features of the classic BRAND PD-Tips.Thanks to patented size coding, HandyStep® touch and HandyStep® electronic from BRAND automatically detect the tip volume. The tips are also compatible with all other instruments of the HandyStep® family and with repetitive pipettes from other manufacturers.The positive displacement principle enables precision work, even with viscous media or media with high vapor pressure. With tightly sealed pistons, the PD-Tips II effectively protect the stepper from aerosols. This prevents contamination and ensures long service life of the instrument.At a glance- Volumes from 0.1 ml to 50 ml- Compatible with different repetitive pipets- Compatible with automatic size detection: HandyStep® touch, HandyStep® electronic, Gilson™ Repetman, Rainin® AutoRep E- Compatible without automatic size detection: HandyStep® S, HandyStep®, [TM="Rainin" AutoRep S, Rainin® AutoRep M, Eppendorf® Multipette® 4780 and EDOS® 5221 (the above-mentioned brands are property of their respective owners)- Cleanroom quality- Comply with the requirements of ISO 8655- Delivered with batch certificate- Manufactured from high-quality raw materials (cylinder: PP / Piston: PE-HD, 0.1 ml: LCP)- Adapter for sizes 25 ml and 50 ml included- PD-Tips II cannot be autoclaved; the adapters of the 25ml and 50ml sizes can be autoclaved at 121° C.Adapters for size 25 and 50 ml PD-Tips, PP, autoclavable. Non-sterile or sterile in BIO-CERT® quality.CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Features and Benefits
  • Dispense with ease – even critical media
  • Save time with automatic size detection
Other Notes
BR702399 (India)
Legal Information
Gilson is a trademark of Gilson, Inc.
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
EDOS is a registered trademark of Eppendorf SE
Eppendorf is a registered trademark of Eppendorf SE
HandyStep is a registered trademark of BRAND GMBH + CO KG
Multipette is a registered trademark of Eppendorf SE
Rainin is a registered trademark of Rainin Instrument, LLC
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z332798BR705710BR705748
description
sterile
description
non-sterile
description
volume 5 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
description
BIO-CERT®, volume 50 mL, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
manufacturer/tradename
BRAND 702399
manufacturer/tradename
BRAND 702398
manufacturer/tradename
BRAND 705710
manufacturer/tradename
BRAND 705748
sterility
sterile
sterility
non-sterile
sterility
non-sterile
sterility
-
material
polypropylene
material
polypropylene
material
high-density polyethylene piston, polypropylene cylinder
material
high-density polyethylene piston, polypropylene cylinder
packaging
pack of 5 ea
packaging
pack of 10 ea
packaging
-
packaging
-
maximum volume
50 mL
maximum volume
50 mL
maximum volume
-
maximum volume
-
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)