Fortfahren mit
Z328839
BRAND® Magnetrührstäbchen, PTFE, zylindrisch
L 5 mm, bar diam. 2 mm
Synonym(e):
PTFE, Stirring rods
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
10 ea
Warenkorb auf Verfügbarkeit prüfen
€ 53,50
Über diesen Artikel
UNSPSC Code:
41103806
NACRES:
NB.72
eCl@ss:
32100100
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137102, BRAND
€ 53,50
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
Quality Segment
material
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137102, BRAND
L
5 mm
bar diam.
2 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137102 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z328650Z328766Z328790
description
L 5 mm, bar diam. 2 mm
description
L 10 mm, bar diam. 3 mm
description
L 20 mm, bar diam. 6 mm
description
L 30 mm, bar diam. 6 mm
manufacturer/tradename
BRAND, BRAND 137102
manufacturer/tradename
BRAND 137108
manufacturer/tradename
BRAND 137115
manufacturer/tradename
BRAND 137125
material
cylindrical , PTFE-coated
material
cylindrical , PTFE-coated
material
PTFE-coated , cylindrical
material
cylindrical , PTFE-coated
Quality Level
200
Quality Level
200
Quality Level
200
Quality Level
200
L
5 mm
L
10 mm
L
20 mm
L
30 mm
bar diam.
2 mm
bar diam.
3 mm
bar diam.
6 mm
bar diam.
6 mm
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)