Merck
TMO169558
Nunc® Gridded Cell Culture Dishes
diam. 60 mm, with lid, Nunclon® Delta treated, case of 400 ea
Synonym(e):
petri dish, treated dish, treated petri dish
Größe auswählen
400 EACH
€ 626,00
€ 626,00
Voraussichtliches Versanddatum28. Mai 2025
Größe auswählen
Ansicht ändern
400 EACH
€ 626,00
About This Item
UNSPSC-Code:
41121812
NACRES:
NB.13
€ 626,00
Voraussichtliches Versanddatum28. Mai 2025
Verpackung
case of 400 ea
Hersteller/Markenname
Nunc 169558
Durchmesser
60 mm
Allgemeine Beschreibung
With wide selections in size and style, Thermo Scientific Nunc Cell Culture Dishes suit a multitude of experimental needs in your cell culture routine. Perform a variety of applications in dishes with the Nunclon Delta surface treatment that promotes maximum adhesion for a broad range of cell types.
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)