Fortfahren mit
TMO167542
Nunc® Edge 2.0 96-well Plate
Nunclon® Delta surface, polystyrene, sterile, with lid, clear, case of 160 ea
Synonym(e):
cell culture plate, cell growth plate, Nunc 167542, cultivation plate, surface treated dish
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
160 ea
Warenkorb auf Verfügbarkeit prüfen
€ 456,00
Über diesen Artikel
NACRES:
NB.15
UNSPSC Code:
41122102
Material:
clear , polystyrene
Sterility:
sterile
Manufacturer/tradename:
Nunc 167542
Packaging:
case of 160 ea
€ 456,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
clear , polystyrene
sterility
sterile
packaging
case of 160 ea
manufacturer/tradename
Nunc 167542
General description
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Use versatile Thermo Scientific Nunc 1.8 mL Externally-threaded Universal Tubes either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.
Features and Benefits
  • The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
  • Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
  • Plates with wells wich are Nunclon™ Delta cell culture surface treated promote cell attachment and growth.
  • Non-treated surface is available for suspension cell culture or general solution-based assays.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
TMO267544TMO267556TMO167554
description
Nunclon® Delta surface, polystyrene, sterile, with lid, clear, case of 160 ea
description
untreated, polystyrene, sterile, with lid, clear, case of 160 ea
description
untreated, polystyrene, sterile, without lid, clear, case of 160 ea
description
Nunclon® Delta surface, polystyrene, sterile, without lid, clear, case of 160 ea
manufacturer/tradename
Nunc 167542
manufacturer/tradename
Nunc 267544
manufacturer/tradename
Nunc 267556
manufacturer/tradename
Nunc 167554
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
clear , polystyrene
material
polystyrene , clear
material
clear , polystyrene
material
polystyrene , clear
packaging
case of 160 ea
packaging
case of 160 ea
packaging
case of 160 ea
packaging
case of 160 ea
Still not finding the right product?
Explore all of our products under Nunc® Edge 2.0 96-well Plate
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)