Fortfahren mit
TMO140410
Nunc® EasyFill Cell Factory System
10 layer, polystyrene, surface area 6320 cm2, sterile, vented, case of 6 ea
Synonym(e):
closed cell system layered storage rack, closed cell system rack, Nunc 140410, cell cultivation system, stacked cell cultivation module
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
6 ea
Warenkorb auf Verfügbarkeit prüfen
€ 2.290,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41161502
Manufacturer/tradename:
Nunc 140410
Feature:
vented
Material:
polystyrene
Packaging:
case of 6 ea
€ 2.290,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polystyrene
sterility
sterile
feature
vented
packaging
case of 6 ea
manufacturer/tradename
Nunc 140410
surface area
6320 cm2
binding type
Nunclon Delta surface
General description
The Nunc® EasyFill Cell Factory systems are a space saving, out-of-the-box, multilayer solution and are as easy to use as a T-flask. Scale up your research and commercial-scale applications with confidence. These systems come with a cerrtified Nunclon® Delta cell culture surface to enable consistent performance from lot to lot and format to format. The 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Features and Benefits
  • "Out of the box" system with no accessories required.
  • Large, vented screw closure for easy handling
  • Versatile port design allows for pouring and aseptic filling.
  • Fill and empty five times faster compared to T175 flasks.
  • Enhances productivity by enabling faster operations.
  • Fast start-up with a simple pour of media and cells into the large opening.
  • Versatile port facilitates pouring and aseptic filling techniques.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
TMO140360TMO140400TMO140250
description
10 layer, polystyrene, surface area 6320 cm2, sterile, vented, case of 6 ea
description
4 layer, polystyrene, surface area 2528 cm2, sterile, vented, case of 4 ea
description
10 layer, polystyrene, surface area 6320 cm2, sterile, vented, case of 2 ea
description
2 layer, polystyrene, surface area 1264 cm2, sterile, vented, case of 6 ea
manufacturer/tradename
Nunc 140410
manufacturer/tradename
Nunc 140360
manufacturer/tradename
Nunc 140400
manufacturer/tradename
Nunc 140250
feature
vented
feature
vented
feature
vented
feature
vented
material
polystyrene
material
polystyrene
material
polystyrene
material
polystyrene
packaging
case of 6 ea
packaging
case of 4 ea
packaging
case of 2 ea
packaging
case of 6 ea
surface area
6320 cm2
surface area
2528 cm2
surface area
6320 cm2
surface area
1264 cm2
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)