Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
10 ea | Warenkorb auf Verfügbarkeit prüfen | € 342,00 |
Über diesen Artikel
€ 342,00
material
cap (Sealing cap), polypropylene , polypropylene copolymer
sterility
non-sterile
feature
RCF 100,605 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3139-0050
nominal capacity
50 mL
General description
Features and Benefits
- Greater chemical resistance than polycarbonate
- Autoclavable
- Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
- 3139-0050 (T2918) can be used up to 100,605 x g in the T29-8x50 rotor
- Comes with polypropylene sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
- Must be filled to 80% of total capacity when centrifuging
- Autoclavable/Leakproof† when used with supplied Nalgene closure; Repeated autoclaving will reduce the life of PC centrifuge tubes.
Packaging
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description nominal capacity 50 mL | description nominal capacity 16 mL | description nominal capacity 30 mL | description nominal capacity 30 mL |
| manufacturer/tradename Nalgene® 3139-0050 | manufacturer/tradename Nalgene® 3139-0016 | manufacturer/tradename Nalgene® 3139-0030 | manufacturer/tradename Nalgene® 3138-0030 |
| sterility non-sterile | sterility non-sterile | sterility - | sterility non-sterile |
| material cap (Sealing cap), polypropylene copolymer | material cap (Sealing cap), polypropylene , polypropylene copolymer , round bottom tube | material cap (Sealing cap), polypropylene , polypropylene copolymer , round bottom tube | material cap (PP sealing cap), round bottom tube, transparent polycarbonate |
| feature RCF 100,605 × g | feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g |
| packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea |
| nominal capacity 50 mL | nominal capacity 16 mL | nominal capacity 30 mL | nominal capacity 30 mL |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge tubes, Oak Ridge Style 3139
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?