Fortfahren mit
SIAL0167
Sigma® cell culture dish
diam. × H 100 mm × 20 mm, tissue-culture treated, sterile, pack of 25 × 20 ea
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121812
sterility
sterile
packaging
pack of 25 × 20 ea
diam. × H
100 mm × 20 mm
surface area
55 cm2
binding type
Tissue Culture (TC)-treated surface
General description
  • Manufactured from optically-clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid supplied with vents to provide consistent gas exchange
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)