Merck
RF353017C-EP
Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges
F-35-30-17, non aerosol tight, with 10 steel sleeves for 15 mL vessels, with 10 adapters for conical tubes and 10 rubber mats for round-bottom tubes
About This Item
UNSPSC-Code:
41103909
Leistungsmerkmale
to hold 20 x 15 conical tubes
to hold 30 x 15 mL glass tubes
fixed angle rotor
non aerosol tight
with 10 steel sleeves for 15 mL vessels
with 10 adapters for conical tubes and 10 rubber mats for round-bottom tubes
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 022639421 (NA)
Eppendorf® 5702705004 (EU)
Parameter
2,750 × g max. RCF
Methode(n)
centrifugation: suitable
Kompatibilität
for use with centrifuge models 5702/5702R/5702RH
Verwandte Kategorien
Allgemeine Beschreibung
Rotors to fit Eppendorf centrifuge models 5702, 5720 R, and 5720 RH
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)