Merck
P9360
Eppendorf® Research® Pro electronic pipette, multichannel
with charging adapter, size 20-300 μL
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Über diesen Artikel
UNSPSC Code:
41121500
Fortfahren mit
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
description
12 channel
feature
electronic
manufacturer/tradename
Eppendorf® 022461508 (USA), Eppendorf® 4860 000.569 (European), Eppendorf® 4860 000.569, Eppendorf® 4860 200.568 (UK)
size
20-300 μL
General description
This pipette provides a wide variety of programmable application parameters, while still remaining simple to use and easy to handle. The styling of the Eppendorf Research pro pipette facilitates fast and accurate performance, with the expected precision and reliability of Eppendorf
• Easy-to-read display screen is set at a 45° angle for easy reference during use
• Motorized piston operation increases reproducibility
• All buttons and controls are large and easily accessible for both right- and left-handed users
• Volume and speed options are controlled with a rocker switch, requiring only a minimal amount of force, avoids potential wrist and hand strain
• Separate tip ejection lever can be adjusted to the most comfortable position for right- and left-handed users
• Fully charged battery performs over 2,000 pipetting cycles reduces downtime
• Automatic shut-off function after 10 minutes of non-use
• Color coded actuate button corresponds with common pipette tip volume sizes
• User-calibratable ideal for use with solutions of different viscosity, density, surface tension, or vapor pressure
• Lower half is removable and autoclavable at 121 °C for simplified decontamination
• Quick recharge NiMH battery-performs up to 200 pipetting strokes after recharging only 30 minutes
• For each program, 10 individual aspirating and dispensing speeds can be programmed
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Eppendorf Research is a registered trademark of Eppendorf SE
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)