Merck
OH30253061
Ohaus® Scout® Portable Touchscreen Balance
Model STX422, weighing capacity 420 g, resolution: 0.01 g, AC/DC input 230 V
Größe auswählen
1 EACH
€ 795,00
€ 795,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EACH
€ 795,00
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.13
€ 795,00
Check Cart for Availability
Materialien
metal weighing plate
Leistungsmerkmale
CE compliant
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30253061
Messbereich Wiegekapazität
420 g, resolution: 0.01 g
Waagschalendurchm.
12 cm
Allgemeine Beschreibung
Ideal for laboratory and industrial applications, the OHAUS Scout features a user-friendly color touchscreen and a compact stackable design. Designed to deliver superior performance with rapid stability testing and high resolution weighing results, these portable scales set a new standard in weighing applications. Their features also include superior overload protection and advanced connectivity options.
Anwendung
Weighing, piece counting,% weighing, check weighing, animal weighing / dynamic weighing, totalizing, density determination, display hold, mole weighing
Leistungsmerkmale und Vorteile
Stability indicator, stackable design with storage cover included (except 1 mg models), mechanical and software overload / underload protection, low battery level indicator, automatic shutdown, auto-tare, slope calibration mass included with models up to 620g.
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)