Fortfahren mit
MTCR1040
MTC Bio Fraction Collector Tube Rack
holds 80 x 1.5 /2.0 mL tubes, assorted colors, pack of 5 ea
Synonym(e):
tube holder, tube storage, tube organizer, tube rack, MTC Bio T1040
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.12
UNSPSC Code:
41122804
Manufacturer/tradename:
MTC Bio R1040
Feature:
holds 80 x 1.5 /2.0 mL tubes
Material:
assorted colors , polypropylene
Packaging:
pack of 5 ea
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
assorted colors , polypropylene
feature
holds 80 x 1.5 /2.0 mL tubes
packaging
pack of 5 ea
manufacturer/tradename
MTC Bio R1040
General description
Tube Rack, fraction collector type, 80 x 1.5mL/2mL, assorted colors (B, G, R, Y, O), pack of 5
Compact, convenient 80 well racks designed to keep your tubes neatly and efficiently organized. These autoclavable polypropylene racks are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification. The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes.
Features and Benefits
  • 80 well (5 rows of 16) for 1.5 to 2.0mL microtubes
  • Rainbow pack available — 5 colors
  • Two frosted marking areas, on the bottom and side
  • Raised numbers beside each well
  • Recessed well bottoms for improved tube stability
Legal Information
MTC is a trademark of MTC Bio
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)