Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 43,40 |
Über diesen Artikel
€ 43,40
material
purple polypropylene (/natural)
feature
to hold 20 x 10-17 mm tubes OR 12 x 25 - 30 mm tubes
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120818
L × W × H
309 mm × 107 mm × 77 mm
well diam. × diam. × diam.
10-13 mm × 13-17 mm × 25-30 mm
General description
Features and Benefits
- Interchangeable tube rack inserts hold multiple tube sizes firmly in place when alternating between sample vessels
- Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube being used
- Open design allows for better filling visibility of tube graduations when filling
- Rack wells have a unique teardrop design that enables better retention of tubes over traditional designs
- Able to rotate the tube rack insert to an angle position to facilitate more comfortable pipetting
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description purple polypropylene (/natural) | description for tubes, 5 ‑ 50 mL, blue, pk of 2 | description blue polypropylene (/natural) | description for tubes, 5 ‑ 50 mL, white, pk of 2 |
| manufacturer/tradename Heathrow Scientific 120818 | manufacturer/tradename Heathrow Scientific HS120187 | manufacturer/tradename Heathrow Scientific 120817 | manufacturer/tradename Heathrow Scientific HS120188 |
| feature to hold 20 x 10-17 mm tubes OR 12 x 25 - 30 mm tubes | feature for tubes 5 ‑ 50 mL, Holds 4 x 12-30 mm tubes, Holds 4 x 5-50 mL tubes | feature to hold 20 x 10-17 mm tubes OR 12 x 25 - 30 mm tubes | feature for tubes 5 ‑ 50 mL, Holds 4 x 12-30 mm tubes, Holds 4 x 5-50 mL tubes |
| material purple polypropylene (/natural) | material blue POM | material blue polypropylene (/natural) | material white POM |
| packaging pkg of 1 ea | packaging pk of 2 | packaging pkg of 1 ea | packaging pk of 2 |
| L × W × H 309 mm × 107 mm × 77 mm | L × W × H 18.1 × 5.6 × 7.6 cm | L × W × H 309 mm × 107 mm × 77 mm | L × W × H 18.1 × 5.6 × 7.6 cm |
| well diam. × diam. × diam. 10-13 mm × 13-17 mm × 25-30 mm | well diam. × diam. × diam. - | well diam. × diam. × diam. 10-13 mm × 13-17 m2 × 25-30 mm | well diam. × diam. × diam. - |
Still not finding the right product?
Explore all of our products under OneRack® Multi
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?