Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 190,00 |
Über diesen Artikel
€ 190,00
material
pink , white
feature
adjustable, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120105
volume measuring range
500-5000 μL
General description
Features and Benefits
- Internal metal parts for increased accuracy and calibration stability, and long life
- Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
- Easy volume adjustment with locking tab to prevent accidental change
- Separate tip ejector
- Large digit volume display
- Color coded and labeled with volume range
- Both top and side have colored coded components for quick visual confirmation
- CE certified and Manufactured in accordance with ISO08655 and GLP regulations
- Autoclavable
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description adjustable, volume 500-5000 μL, pink, single-channel | description adjustable, volume 0.1-2 μL, green, single-channel | description adjustable, volume 1000-10000 μL, red, single-channel | description adjustable, volume 0.5-10 μL, light blue, single-channel |
| material pink , white | material green , white | material red , white | material white , light blue |
| volume measuring range 500-5000 μL | volume measuring range 0.1-2 μL | volume measuring range 1000-10000 μL | volume measuring range 0.5-10 μL |
| manufacturer/tradename Heathrow Scientific HS120105 | manufacturer/tradename Heathrow Scientific HS120097 | manufacturer/tradename Heathrow Scientific HS120106 | manufacturer/tradename Heathrow Scientific HS120098 |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| feature adjustable, single-channel | feature adjustable, single-channel | feature adjustable, single-channel | feature adjustable, single-channel |
Still not finding the right product?
Explore all of our products under Pearl™ Pipette
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?