Fortfahren mit
HS120044
Microtube Storage Box
Holds 100 x 1.5-2.0 mL tubes, assorted colors polypropylene ((blue, green, purple, yellow, orange), pk of 5
Synonym(e):
tube rack, tube storage
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.12
UNSPSC Code:
41123300
Manufacturer/tradename:
Heathrow Scientific HS120044
Feature:
Holds 100 x 1.5-2.0 mL tubes
Material:
assorted colors polypropylene ((blue, green, purple, yellow, orange)
Packaging:
pk of 5
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
assorted colors polypropylene ((blue, green, purple, yellow, orange)
feature
Holds 100 x 1.5-2.0 mL tubes
packaging
pk of 5
manufacturer/tradename
Heathrow Scientific HS120044
L × W × H
5.6 in. × 6 in. × 2.2 in.
well diam.
12.3 mm
Features and Benefits
  • for larger storage requirements of 1.5-2.0 mL microtubes or cryogenic vials
  • easy-open hinged lid with snap clasp
  • autoclavable
  • designed for quick visual identification
  • uniform wells with location indentification
  • tube capacity is indicated on the outside of the box
  • translucent vibrant colors with frosted labeling area
  • debossed with a distinctive wave design
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
HS120201R5651R5526
description
Holds 100 x 1.5-2.0 mL tubes, assorted colors polypropylene ((blue, green, purple, yellow, orange), pk of 5
description
blue polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
description
assorted colors
description
orange
manufacturer/tradename
Heathrow Scientific HS120044
manufacturer/tradename
Heathrow Scientific HS120201
manufacturer/tradename
Heathrow Scientific HS29022G
manufacturer/tradename
Heathrow Scientific HS29022F
feature
Holds 100 x 1.5-2.0 mL tubes
feature
Holds 100 x 1.5-2.0 mL tubes
feature
Holds 12 x 15 mL tubes, Holds 32 x 0.5 mL tubes, Holds 32 x 1.5-2 mL tubes, Holds 4 x 50 mL tubes
feature
Holds 12 x 15 mL tubes, Holds 32 x 0.5 mL tubes, Holds 32 x 1.5-2 mL tubes, Holds 4 x 50 mL tubes
material
assorted colors polypropylene ((blue, green, purple, yellow, orange)
material
blue polypropylene
material
polypropylene (heavy duty)
material
polypropylene (heavy duty)
packaging
pk of 5
packaging
pk of 5
packaging
-
packaging
-
well diam.
12.3 mm
well diam.
12.3 mm
well diam.
-
well diam.
-
L × W × H
5.6 in. × 6 in. × 2.2 in.
L × W × H
-
L × W × H
-
L × W × H
-
Still not finding the right product?
Explore all of our products under Microtube Storage Box
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)