Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
natural polypropylene
feature
Holds 100 x 1.5-2.0 mL tubes
packaging
pk of 5
manufacturer/tradename
Heathrow Scientific HS120042
L × W × H
5.6 in. × 6 in. × 2.2 in.
size
100 wells
well diam.
12.3 mm
Features and Benefits
- for larger storage requirements of 1.5-2.0 mL microtubes or cryogenic vials
- easy-open hinged lid with snap clasp
- autoclavable
- designed for quick visual identification
- uniform wells with location indentification
- tube capacity is indicated on the outside of the box
- translucent vibrant colors with frosted labeling area
- debossed with a distinctive wave design
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Holds 100 x 1.5-2.0 mL tubes, size 100 wells, natural polypropylene, pk of 5 | description Holds 100 x 1.5-2.0 mL tubes, black polypropylene, pk of 5 | description orange polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes | description green polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes |
| manufacturer/tradename Heathrow Scientific HS120042 | manufacturer/tradename Heathrow Scientific HS120043 | manufacturer/tradename Heathrow Scientific HS120204 | manufacturer/tradename Heathrow Scientific HS120202 |
| feature Holds 100 x 1.5-2.0 mL tubes | feature Holds 100 x 1.5-2.0 mL tubes | feature Holds 100 x 1.5-2.0 mL tubes | feature Holds 100 x 1.5-2.0 mL tubes |
| material natural polypropylene | material black polypropylene | material orange polypropylene | material green polypropylene |
| packaging pk of 5 | packaging pk of 5 | packaging pk of 5 | packaging pk of 5 |
| L × W × H 5.6 in. × 6 in. × 2.2 in. | L × W × H 5.6 in. × 6 in. × 2.2 in. | L × W × H - | L × W × H - |
| size 100 wells | size - | size - | size - |
Still not finding the right product?
Explore all of our products under Microtube Storage Box
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?