Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
feature
Holds 12 x pipettors
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120026
size
332x193 mm
color
blue
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description blue | description for 6 Eppendorf Xplorer® or Xplorer® plus, AC/DC input 100 V / 240 V, 50/60 Hz, includes four interchangeable power plugs | description blue magnetic rack | description purple to clear, 8 of each color for top + 8 clear bottoms |
| feature Holds 12 x pipettors | feature Holds 6 x pipetors, includes four interchangeable power plugs | feature Holds 0.1 to 50 mL pipettes or cans | feature Holds 16 x pipettors |
| manufacturer/tradename Heathrow Scientific HS120026 | manufacturer/tradename Eppendorf® 3116000023 (Global) | manufacturer/tradename Heathrow Scientific HS20612M | manufacturer/tradename Heathrow Scientific HS120481 |
| packaging pack of 1 ea | packaging - | packaging - | packaging - |
| size 332x193 mm | size - | size - | size - |
| color blue | color - | color - | color purple to clear, 8 of each color for top + 8 clear bottoms |
Still not finding the right product?
Explore all of our products under Flip & Grip® pipette holder or pipette stand
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?