Fortfahren mit
EPS44I211005
Eppendorf® Innova® S44i Shaker
refrigerated, photosythetic LED light bank, 1" orbit, AC/DC input 120 V
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41103811
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
feature
photosythetic LED light bank, 1" orbit, refrigerated
manufacturer/tradename
Eppendorf® S44I211005
General description
The Innova S44i is a large capacity, temperature controlled (refrigerated) stackable shaker allowing the use of high-volume flasks in large numbers in a compact footprint. The iconic slide up door provides easy access to your samples. The Eppendorf X-Drive allows for shaking high loads at high speeds – even when stacked. When there is an instance of an unbalanced load, the X-Drive′s counterbalance adjustment allows for smooth shaking. The optional photosynthetic LED lightbank provides the ability to evenly light the entire platform across a wide intensity range – and it′s programmable. The VisioNize touch interface introduces onboard monitoring, user management and the ability to export data via USB; or network integration via ethernet. A comprehensive service offering including IQ/OQ is also available.
Product Information
The Innova S44i is a large capacity, temperature controlled (refrigerated) stackable shaker allowing the use of high-volume flasks in large numbers in a compact footprint. The iconic slide up door provides easy access to your samples. The Eppendorf X-Drive allows for shaking high loads at high speeds – even when stacked. When there is an instance of an unbalanced load, the X-Drive′s counterbalance adjustment allows for smooth shaking. The optional photosynthetic LED lightbank provides the ability to evenly light the entire platform across a wide intensity range – and it′s programmable. The VisioNize touch interface introduces onboard monitoring, user management and the ability to export data via USB; or network integration via ethernet. A comprehensive service offering including IQ/OQ is also available.
Maximum capacity
  • The Innova S44i has optimized chamber, platform, and features to maximize your options and capacity.
  • Up to 136 % more capacity than the previous Innova 44 with a smaller footprint
  • Up to 102 % higher flask capacity than competitor
  • Double- or triple-stack the Innova S44i to increase capacity without sacrificing more valuable lab space
Eppendorf X-Drive
  • The Eppendorf X-Drive is precisely engineered with five-eccentric shafts to balance even the heaviest load while providing consistent and smooth shaking.
  • The intelligent counterbalance technology of the Eppendorf X-Drive ensures vibration-free shaking even with uneven loads. This advanced technology senses imbalance and decelerates to a safe speed to protect you and your cultures.
Advanced control
The new touchscreen user interface of the Innova S44i offers more than just an easy operation. This multi-touch capacitive display makes all the information you need
easy to monitor and track.
  • Information is just one touch away
  • Intuitive operating, distinct design
  • Manage multiple users with different levels of authorization
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Innova is a registered trademark of Eppendorf, Inc. 
Still not finding the right product?
Explore all of our products under Eppendorf® Innova® S44i Shaker
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)