Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NB.73
UNSPSC Code:
41122804
eCl@ss:
32032201
Manufacturer/tradename:
Eppendorf® M1289-0100 (Global)
Feature:
holds 80 tubes
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltensize
large
feature
holds 80 tubes
manufacturer/tradename
Eppendorf® M1289-0100 (Global)
size
450x80x129 mm
General description
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Application
Securly stores tubes in Eppendorf Innova and Excella shakers.
Features and Benefits
- Adjustable angle and tube height support for maximun flexibility
- Inserts help retain tubes to prevent spinning in the rack
- Finger-tight knobs for quick release of rack from the base plate for easy transportation to the bench
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Excella is a registered trademark of Eppendorf, Inc.
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf SE
Still not finding the right product?
Explore all of our products under Test Tube Rack, for Eppendorf® New Brunswick™ Innova® and Eppendorf® Excella® Shakers
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?