Merck
EP4924000924
Eppendorf® Reference® 2 G
pack of 3, Option 3, 100-1000 uL/0.5-5 mL/1-10 mL, includes epT.I.P.S.® box
Größe auswählen
3 EA
€ 900,00
€ 900,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
3 EA
€ 900,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 900,00
Check Cart for Availability
Leistungsmerkmale
single-channel
Option 3, 100-1000 uL/0.5-5 mL/1-10 mL, includes epT.I.P.S.® box
Verpackung
pack of 3
Hersteller/Markenname
Eppendorf® 4924000924
Volumen
× 10 mL
× 100 μL
× 5 mL
Verwandte Kategorien
Allgemeine Beschreibung
Extraordinary precision and accuracy – Eppendorf´s premium pipette
Besides improved ergonomics the design focus lays on providing the most accurate results possible, optimum user safety, reliability and robustness for a long service life. The innovative single button operation is not only fast and easy, but it also allows active aerosol1) reduction and thus protects the user, the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility, Eppendorf′s Reference 2 is an ideal instrument for these liquid handling applications.
Single-Button Operation
  • Lower Operating forces for aspiration/dispensing/tip ejection limit strain on the user.
  • Allows users to move more quickly through their pipetting tasks by not having to move the thumb from side to side to eject tips.
  • With the ability to eject a tip immediately following dispensing, aerosol contamination of your pipette is considerably reduced.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)