Merck
EP3124000059
Eppendorf Research® plus Pipette, fixed volume
single-channel, volume 25 μL, yellow
Größe auswählen
1 EA
€ 199,00
€ 199,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 199,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 199,00
Check Cart for Availability
Materialien
yellow
Leistungsmerkmale
single-channel
mechanical
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 3121000058 (Global)
Volumen
25 μL
Allgemeine Beschreibung
The Eppendorf Research plus mechanical pipette is the result of over 60 years of experience in liquid handling and is one of the most frequently used pipettes in the world. Thanks to the groundbreaking Eppendorf PhysioCare Concept, the Research plus is very ergonomic and protects your health in the laboratory. It is characterized by low weight and low operating forces, is extremely reliable, fully autoclavable and easy to use. It feels good to know that you are doing the right thing – knowing that you are working with one of the most well-known and advanced pipettes in the world. A spring-loaded tip cone, an option for temporary adjustment, an improved volume display – and all this in a lightweight, fully autoclavable pipette: that is the Eppendorf Research plus manual pipette. In 2021, the Research plus single-channel variable volume pipettes were certified with the ACT Environmental Impact Factor Label by the non-profit organization My Green Lab . This makes them a good choice for laboratories looking for more sustainable products.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)