Fortfahren mit
CLS6710
Corning® LSE Multiposition Digital Vortexer
AC/DC input 230 V, AC/DC input 50 Hz, AC/DC input, Schuko plug, AC/DC input, UK plug
Synonym(e):
shaker
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 1.440,00
Über diesen Artikel
NACRES:
NB.02
UNSPSC Code:
41103814
€ 1.440,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
manufacturer/tradename
Corning 6710
6710
chamber W × D × H
(10.3 in.) (13.3 in.) (5.1 in.)
General description
The Corning Digital Vortexer is a multipurpose shaker dedicated to a variety of shaking and mixing applications. Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes. The microplate platform is designed to securely hold up to four microplates. Operation of the vortexer can be timed or continuous, while the speed is variable. The broad speed range allows the vortexer to be used for gentle mixing as well as vigorous agitation. A powerful motor, and a chemically resistant and easy to clean surface makes the equipment a perfect choice for years of trouble-free use.
Features and Benefits
  • Three versatile platform options (available separately)
  • Compact design with easy-access control panel
  • Non-slip rubber mat to keep containers in place
  • Safe for cold room or incubator use
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS6777CLS6776CLS6781NP
description
AC/DC input 230 V, AC/DC input 50 Hz, AC/DC input, Schuko plug, AC/DC input, UK plug
description
vortex mixer w/ standard tube head, 230 V, UK plug, 1/cs
description
vortex mixer w/ standard tube head, 230 V, EU plug, 1/cs
description
orbital shaker, without platform, 230V, EU plug, 1/cs
chamber W × D × H
(10.3 in.) (13.3 in.) (5.1 in.)
chamber W × D × H
-
chamber W × D × H
-
chamber W × D × H
-
manufacturer/tradename
Corning 6710
6710
manufacturer/tradename
Corning 6777
manufacturer/tradename
Corning 6776
manufacturer/tradename
Corning 6781-NP
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)