Fortfahren mit
CLS6704
Corning® LSE Platform Rocker
AC/DC input 230 V, Corning 6704, AC/DC input 50/60 Hz, AC/DC input, Schuko plug
Synonym(e):
equipment replacement accessory
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 1.050,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41103811
€ 1.050,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
AC/DC input
, UK plug
manufacturer/tradename
Corning 6704
General description
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications. The adjustable rocking speed and fixed 7 degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots.
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations.
Features and Benefits
  • Low profile and compact size
  • 11-pound load capacity
  • Non-slip rubber pads to prevent sample containers from moving during operation
  • Double platform model available for increased capacity
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS6706Z755567Z755559
description
AC/DC input 230 V, Corning 6704, AC/DC input 50/60 Hz, AC/DC input, Schuko plug
description
Double Platform, AC/DC input 230 V, AC/DC input 50/60 Hz, AC/DC input, Schuko plug
description
AC/DC input 230 V AC, UK plug, CE compliant
description
AC/DC input 230 V AC, European 2-pin plug, CE compliant
AC/DC input
, UK plug
AC/DC input
-
AC/DC input
-
AC/DC input
-
manufacturer/tradename
Corning 6704
manufacturer/tradename
Corning 6706
manufacturer/tradename
Benchmark Scientific B3D1308-230-UK plug
manufacturer/tradename
Benchmark Scientific B3D1308-230-EU plug
Still not finding the right product?
Explore all of our products under Corning® LSE Platform Rocker
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)