Merck
CLS4985P250
Corning® narrow mouth Erlenmeyer flask, reusable
capacity 250 mL, polypropylene, with GL-52 PP screw cap
Synonym(e):
conical flasks, laboratory flasks
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
narrow-mouth
polypropylene
screw top cap
screw top cap
Sterilität
non-sterile
Leistungsmerkmale
cap
graduated 10 mL (intervals)
Verpackung
pack of 6 ea
Hersteller/Markenname
Corning 4985P-250
Kapazität
250 mL
Allgemeine Beschreibung
Translucent polypropylene flasks are manufactured with blue graduations for reference. The flasks have a screw neck and are supplied with a polypropylene cap.
Sonstige Hinweise
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.