Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1000 ea | Warenkorb auf Verfügbarkeit prüfen | € 337,00 |
Über diesen Artikel
UNSPSC Code:
41121500
NACRES:
NB.01
eCl@ss:
32070201
Capacity:
1 mL
Material:
polystyrene
Packaging:
pkg of (50 per bag)
€ 337,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polystyrene
sterility
sterile
manufacturer/tradename
Corning 4975
packaging
pkg of (50 per bag)
capacity
1 mL
volume
1 mL
General description
- Corning® aspirating pipettes are made of optically clear polystyrene for durability and good mechanical strength.
- The sterile, ungraduated, and unplugged pipettes are best suited for aspirating liquid aseptically using vacuum suction.
- Individually wrapped and bulk packed.
- Disposable and certified non-pyrogenic.
Aspirating pipettes are sterile, ungraduated and unplugged polystyrene pipettes for aspirating liquid using vacuum suction.
Legal Information
Corning is a registered trademark of Corning, Inc.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description volume 1 mL | description volume 5 mL | description volume 2 mL, polystyrene, plain, paper/plastic wrapped | description volume 2 mL |
| manufacturer/tradename Corning 4975 | manufacturer/tradename Corning 9099 | manufacturer/tradename Corning 9016 | manufacturer/tradename Corning 9186 |
| capacity 1 mL | capacity 5 mL | capacity 2 mL | capacity 2 mL |
| material polystyrene | material polystyrene | material polystyrene | material polystyrene |
| packaging pkg of (50 per bag) | packaging case of 200 ea, pack of 50 ea | packaging pack of 50 ea, case of 500 ea | packaging pack of 100 ea, case of 1000 ea |
| volume 1 mL | volume 5 mL | volume 2 mL | volume 2 mL |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
Still not finding the right product?
Explore all of our products under Corning® aspiration pipettes
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?