Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
100 ea | Warenkorb auf Verfügbarkeit prüfen | € 419,00 |
Über diesen Artikel
€ 419,00
material
U-bottom bottom, white polystyrene
sterility
sterile
packaging
pack of 100 ea (Individually wrapped)
manufacturer/tradename
Corning 4873
capacity
100 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description white polystyrene, capacity 100 mL, pack of 100 ea (Individually wrapped) | description white polystyrene, capacity 100 mL, pack of 200 ea (Bulk) | description white polystyrene, capacity 50 mL | description capacity 25 mL, individually wrapped, sterile |
| capacity 100 mL | capacity 100 mL | capacity 50 mL | capacity 25 mL |
| manufacturer/tradename Corning 4873 | manufacturer/tradename Corning 4872 | manufacturer/tradename Corning 4871 | manufacturer/tradename Axygen Inc, Corning |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material U-bottom bottom, white polystyrene | material U-bottom bottom, white polystyrene | material U-bottom bottom, white polystyrene | material - |
| packaging pack of 100 ea (Individually wrapped) | packaging pack of 200 ea (Bulk) | packaging pack of 100 ea (Individually wrapped) | packaging - |
Still not finding the right product?
Explore all of our products under Corning® Costar® Reagenzien-Reservoirs
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?