Merck
CLS451515L
Corning® direct overhead drive assembly for ProCulture® spinner flasks
paddle assembly for 15L Proculture spinner flask w/ direct drive overhead motor, 1/cs
Synonym(e):
Corning container
Größe auswählen
1 EA
€ 421,00
€ 421,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 421,00
About This Item
UNSPSC-Code:
41122104
NACRES:
NB.43
€ 421,00
Check Cart for Availability
Sterilität
non-sterile
Verpackung
case of 1
pack of 1
Hersteller/Markenname
Corning 4515-15L
Allgemeine Beschreibung
Direct drive paddle assembly for use with direct drive overhead motors on series 4510 and 4512, 15L Proculture® Spinner Flasks. The paddle assemblies can be used with either 100 mm or 120 mm direct drive center cap assemblies.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)