Fortfahren mit
CLS4510
Corning® 384 well microplate, low volume
High Binding surface treatment, black polystyrene, round bottom, non-sterile, lid: no, pack of 10
Synonym(e):
384 multiwell plates
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.24
UNSPSC Code:
41121800
Material:
black bottom, black polystyrene , round bottom
Size:
384 wells
Sterility:
non-sterile
Binding type:
high binding surface
Feature:
lid: no, skirt, High Binding surface treatment
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
black bottom, black polystyrene , round bottom
sterility
non-sterile
feature
lid: no, skirt, High Binding surface treatment
packaging
pack of 10, case of 50
manufacturer/tradename
Corning 4510
size
384 wells
well volume
35 μL
wells
384 wells , low volume
working volume
2-20 μL
binding type
high binding surface
Legal Information
Corning is a registered trademark of Corning, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS3822CLS4511CLS4512
description
High Binding surface treatment, black polystyrene, round bottom, non-sterile, lid: no, pack of 10
description
Tissue Culture (TC)-treated surface, black polystyrene, round bottom, sterile, lid, pack of 10
description
black polystyrene, round bottom, non-sterile, lid: no, non-treated, pack of 10
description
white polystyrene, round bottom, non-sterile, lid: no, pack of 10
feature
High Binding surface treatment, lid: no, skirt
feature
skirt, lid
feature
lid: no, non-treated, skirt
feature
non-treated, lid: no, skirt
manufacturer/tradename
Corning 4510
manufacturer/tradename
Corning 3822
manufacturer/tradename
Corning 4511
manufacturer/tradename
Corning 4512
sterility
non-sterile
sterility
sterile
sterility
non-sterile
sterility
non-sterile
binding type
high binding surface
binding type
Tissue Culture (TC)-treated surface
binding type
non-treated surface
binding type
non-treated surface
material
black bottom, round bottom , black polystyrene
material
black polystyrene , round bottom , black bottom
material
round bottom , black polystyrene , black bottom
material
round bottom , white bottom, white polystyrene , round bottom white
size
384 wells
size
384 wells
size
384 wells
size
384 wells
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)