Über diesen Artikel
NACRES:
NB.73
UNSPSC Code:
41122800
Manufacturer/tradename:
Corning 440129
sterility
non-sterile
manufacturer/tradename
Corning 440129
diam. × L
5/16 × 18 in. , two-pieces
General description
Corning offers a stainless steel, two piece, 18 in. (45.7cm) support rod with a 5/16 in. (0.8 cm) diameter that consists of two 9 in. rods that screw together. It screws into the base of Corning hot plate and stirrer models PC-400/410/420, PC-400D/410D/420D and PC-600D/610D/620D.
Legal Information
Corning is a registered trademark of Corning, Inc.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description diam. × L 5/16 × 18 in. , two-pieces | description stainless steel | description AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 5 in. × 7 in., CE compliant | description AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 10 in. × 10 in., CE compliant |
| manufacturer/tradename Corning 440129 | manufacturer/tradename Corning 400084 | manufacturer/tradename Corning 6796-420D | manufacturer/tradename Corning 6796-620D |
| sterility non-sterile | sterility - | sterility - | sterility - |
| diam. × L 5/16 × 18 in. , two-pieces | diam. × L 5/16 in. × 12 in. | diam. × L - | diam. × L - |
Still not finding the right product?
Explore all of our products under Support Rod for Corning® hot plates and stirrers
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?