Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41121812
Material:
polystyrene , round
Sterility:
sterile
Manufacturer/tradename:
Corning 430591
Packaging:
sleeve of 20
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polystyrene , round
sterility
sterile
packaging
sleeve of 20
manufacturer/tradename
Corning 430591
technique(s)
cell culture: suitable
D × H
100 mm × 20 mm
surface area
55 cm2
General description
- Manufactured from optically-clear virgin polystyrene
- Not treated for cell attachment, for applications where cell attachment is not desired
- Sterilized by gamma radiation and certified nonpyrogenic
- Have stacking beads to aid in handling
- Supplied with vents to provide consistent gas exchange
Legal Information
Corning is a registered trademark of Corning, Inc.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description D × H 100 mm × 20 mm | description D × H 60 mm × 15 mm | description D × H 35 mm × 10 mm | description D × H 150 mm × 25 mm |
| manufacturer/tradename Corning 430591 | manufacturer/tradename Corning 430589 | manufacturer/tradename Corning 430165 | manufacturer/tradename Corning 430597 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material polystyrene , round | material round , polystyrene | material polystyrene , round | material polystyrene , round |
| packaging sleeve of 20 | packaging sleeve of 20 | packaging sleeve of 20 | packaging sleeve of 5 |
| surface area 55 cm2 | surface area 21 cm2 | surface area 8 cm2 | surface area 148 cm2 |
| D × H 100 mm × 20 mm | D × H 60 mm × 15 mm | D × H 35 mm × 10 mm | D × H 150 mm × 25 mm |
Still not finding the right product?
Explore all of our products under Corning® non-treated culture dishes
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?