Fortfahren mit
CLS3829
Corning® multicoated microplates
well size 1536, black polystyrene (low base), flat bottom wells clear, non-sterile, lid: no
Synonym(e):
3829, multicoated
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
10 ea
Warenkorb auf Verfügbarkeit prüfen
€ 2.100,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41122102
Material:
black polystyrene (low base), flat bottom wells clear , square wells
Size:
1536 wells
€ 2.100,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
black polystyrene (low base), flat bottom wells clear , square wells
sterility
non-sterile
feature
lid: no
manufacturer/tradename
Corning 3829
size
1536 wells
well size
1536
well volume
12.5 μL
well working volume
up to 8 μL
suitability
suitable for (cell cuture)
General description
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Low base designed for optimal bottom reader sensitivity
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers
Legal Information
Corning is a registered trademark of Corning, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS3651CLS3640CLS3679
description
well size 1536, black polystyrene (low base), flat bottom wells clear, non-sterile, lid: no
description
-
description
surface treatment, Non-binding Surface, clear polystyrene, flat bottom, non-sterile, lid: no, pack of 25
description
half area, flat bottom clear, pkg of 25 per bag, non-sterile, lid: no
size
1536 wells
size
96 wells
size
384 wells
size
96 wells
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
material
square wells, black polystyrene (low base), flat bottom wells clear
material
polystyrene , flat bottom wells clear wells, round clear wells, black polystyrene plate
material
polystyrene , flat bottom , square clear wells, flat bottom wells clear wells, clear polystyrene
material
round clear wells, clear polystyrene plate, flat bottom clear
well size
1536
well size
-
well size
-
well size
-
well working volume
up to 8 μL
well working volume
-
well working volume
-
well working volume
25-125 μL
manufacturer/tradename
Corning 3829
manufacturer/tradename
Corning 3651
manufacturer/tradename
Corning 3640
manufacturer/tradename
Corning 3679
Still not finding the right product?
Explore all of our products under Corning® multicoated microplates
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)