Fortfahren mit
CLS3775
Corning® CrystalEX 384 well protein crystallization plates
cyclic olefin copolymer (COC), flat bottom, non-sterile, lid: no, pack of 10
Synonym(e):
384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well protein crystallization plates, protein crystallization plates
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
50 ea
Warenkorb auf Verfügbarkeit prüfen
€ 1.460,00
Über diesen Artikel
NACRES:
NB.15
UNSPSC Code:
41121800
Material:
clear cyclic olefin copolymer (COC) plate, cyclic olefin copolymer (COC) , flat bottom
Size:
384 wells
€ 1.460,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
clear cyclic olefin copolymer (COC) plate, cyclic olefin copolymer (COC) , flat bottom
sterility
non-sterile
feature
lid: no, plate format: 384 well standard (1 protein well/reservoir well)
packaging
pack of 10, case of 50
manufacturer/tradename
Corning 3775
reservoir volume
105 μL
size
384 wells
well volume
4 μL , protein well volume
suitability
suitable for (cell cuture)
General description
Corning® CrystalEX 384 Well Protein Crystallization Plates
  • 384 well microplate designed for full automation in crystal screening, built to meet industry standards for 384 well microplate footprint and well locations
  • Features 192 reservoir wells (105 μL/well) and 192 corresponding protein wells (4 μL/well)
  • Reservoir wells and protein wells are positioned to be compatible with multihead dispensing equipment (up to 96 well heads)
  • Manufactured from an advanced polymer with high resistance to commonly used solvents, including acetone, acetic acid, butanone, ethanol, isopropanol, methanol, DMSO, nitric acid (65%), sulfuric acid (40%), hydrochloric acid (36%), and ammonia solution (33%)
  • Low background polarization and high optical clarity allows protein crystals to be easily viewed under polarized light with minimal background interference
  • Low water absorption of the polymer prevents loss of protein drop volume
Legal Information
Corning is a registered trademark of Corning, Inc.
CrystalEX is a trademark of Corning, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS3550CLS3552CLS3553
description
cyclic olefin copolymer (COC), flat bottom, non-sterile, lid: no, pack of 10
description
COC, non-treated, 3:1 1uL conical flat bottom crystal cup wells, clear, nonsterile
description
COC, non-treated, 3:1 2uL round bottom wells, clear, nonsterile
description
COC, non-treated, 3:1 2uL conical flat bottom wells, clear, nonsterile
size
384 wells
size
96 wells
size
96 wells
size
96 wells
suitability
suitable for (cell cuture)
suitability
suitable for (sitting drop vapor diffusion high-throughput (HT) protein crystallization)
suitability
suitable for (sitting drop vapor diffusion high-throughput (HT) protein crystallization)
suitability
suitable for (sitting drop vapor diffusion high-throughput (HT) protein crystallization)
material
clear cyclic olefin copolymer (COC) plate, flat bottom
material
clear plate, conical bottom clear wells, cyclic olefin copolymer (COC)
material
clear plate, cyclic olefin copolymer (COC) , round bottom clear wells
material
clear plate, conical bottom clear wells, cyclic olefin copolymer (COC)
packaging
pack of 10, case of 50
packaging
pack of 10 ea, case of 50 ea
packaging
pack of 10 ea, case of 50 ea
packaging
pack of 10 ea, case of 50 ea
reservoir volume
105 μL
reservoir volume
100 μL
reservoir volume
100 μL
reservoir volume
100 μL
manufacturer/tradename
Corning 3775
manufacturer/tradename
Corning 3550
manufacturer/tradename
Corning 3552
manufacturer/tradename
Corning 3553
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)