Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
black polystyrene , clear bottom, flat bottom , polystyrene
sterility
non-sterile
feature
lid: no, skirt
packaging
case of 100 ea, pack of 25
manufacturer/tradename
Corning 3762
maximum volume
112 μL
size
384 wells
working volume
80 μL
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
- Corning® 384 well microplate, low flange black polystyrene, flat bottom, non-sterile, with no lid conforms to standard microplate footprint and dimensions.
- Untreated (or medium binding) polystyrene surface is hydrophobic.
- Applicable for fluorescent assays using either top or bottom detection plate readers.
- Recommended working volume of up to 20 to 80 μL.
- Binding capacity of approximately 100 to 200ng IgG/cm².
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description black polystyrene, flat bottom, non-sterile, lid: no, pack of 25 | description Tissue Culture (TC)-treated surface, white polystyrene, flat bottom, sterile, lid, pack of 10 | description high binding surface, white polystyrene, flat bottom, non-sterile, lid: no, pack of 50 | description black polystyrene, flat bottom, non-sterile, lid: no, pack of 50 |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| size 384 wells | size 384 wells | size 384 wells | size 384 wells |
| material polystyrene , clear bottom, black polystyrene , flat bottom | material polystyrene , white polystyrene , square white wells, white bottom, flat bottom | material square white wells, white polystyrene , flat bottom , white bottom | material polystyrene , flat bottom , square black wells, black bottom, black polystyrene |
| feature lid: no, skirt | feature lid, skirt | feature plate format: 384 well standard, skirt, lid: no | feature lid: no, skirt |
| working volume 80 μL | working volume 80 μL | working volume 80 μL | working volume 80 μL |
| binding type non-treated surface | binding type Tissue Culture (TC)-treated surface | binding type high binding surface | binding type Tissue Culture (TC)-treated surface |
Still not finding the right product?
Explore all of our products under Corning® 384 well microplate, low flange
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?