Fortfahren mit
Vero-Hektor
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41106514
Biological source:
African green monkey kidney
Growth mode:
Adherent
Karyotype:
Not Specified
Morphology:
Fibroblast-like
Manufacturer/tradename:
ECACC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
biological source
African green monkey kidney
manufacturer/tradename
ECACC
packaging
tube of 5 μg 03092503-DNA-5UG, pkg of vial of cells 03092503-1VL
growth mode
Adherent
karyotype
Not Specified
morphology
Fibroblast-like
receptors
Not specified
technique(s)
cell culture: suitable using | mammalian
Biochem/physiol Actions
African green monkey kidney cells, serum-free
Vero cell line (Sigma catalog no. 84113001) adapted to grow in the chemically defined protein- and peptide-free medium Hektor G (Cell Culture Technologies). This cell line is substrate dependent, i.e., adherent and may be suitable for the replication of viral particles.
Preparation Note
Hektor G medium (Cell Culture Technologies http://www.cellculture.com/) + 4 mM L-Glutamine (G7513). When freezing cells down use Hektor G medium + 4 mM L-Glutamine (G7513) + 10% DMSO (D2650).
Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 1-3x10,000 cells/cm2. Use EDTA only (no trypsin); 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)