Merck
C9901
N6-Cyclohexyladenosine
Synonym(e):
CHA
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
Empirische Formel (Hill-System):
C16H23N5O4
CAS-Nummer:
Molekulargewicht:
349.38
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Fortfahren mit
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
InChI key
SZBULDQSDUXAPJ-XNIJJKJLSA-N
SMILES string
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NC4CCCCC4)ncnc23
InChI
1S/C16H23N5O4/c22-6-10-12(23)13(24)16(25-10)21-8-19-11-14(17-7-18-15(11)21)20-9-4-2-1-3-5-9/h7-10,12-13,16,22-24H,1-6H2,(H,17,18,20)/t10-,12-,13-,16-/m1/s1
form
powder
storage temp.
2-8°C
Quality Level
Gene Information
Suchen Sie nach ähnlichen Produkten? Aufrufen Leitfaden zum Produktvergleich
Biochem/physiol Actions
Selective A1 adenosine receptor agonist.
Features and Benefits
This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Lagerklasse
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
H Fraser et al.
British journal of pharmacology, 128(1), 197-205 (1999-09-28)
1. Cardioprotection by adenosine A1 receptor activation limits infarct size and improves post-ischaemic mechanical function. The mechanisms responsible are unclear but may involve alterations in myocardial glucose metabolism. 2. Since glycogen is an important source of glucose during ischaemia, we
Tyson B Brust et al.
Neuropharmacology, 53(8), 906-917 (2007-10-31)
Adenosine A1 receptors are ubiquitous mediators of presynaptic inhibition of neurotransmission in the central nervous system, yet the signalling pathway linking A1 receptor activation and decreased neurotransmitter release remains poorly resolved. We tested the contribution of c-Jun N-terminal kinase (JNK)
Seiji Miyazawa et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 295(3), R991-R996 (2008-07-04)
Body temperature drops dramatically during hibernation, but the heart retains the ability to contract and is resistant to induction of arrhythmia. Although adaptive changes in the heart prior to hibernation may be involved in the cold-resistant property, it remains unclear
Domenico Tupone et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(36), 14512-14525 (2013-09-06)
Since central activation of A1 adenosine receptors (A1ARs) plays an important role in the induction of the hypothermic and hypometabolic torpid state in hibernating mammals, we investigated the potential for the A1AR agonist N6-cyclohexyladenosine to induce a hypothermic, torpor-like state
Gerard J Marek
Neuropharmacology, 56(8), 1082-1087 (2009-03-28)
Modulation of glutamatergic neurotransmission by metabotropic glutamate2/3 (mGlu2/3) receptor agonists effectively treats seemingly diverse neuropsychiatric illness such as generalized anxiety disorder and schizophrenia. Activation of adenosine A(1) heteroceptors, like mGlu2 autoreceptors, decreases glutamate release in the medial prefrontal cortex (mPFC)