Merck
30-2490
Sigma-Aldrich
Zinn(II)-chlorid Dihydrat
SAJ first grade, ≥90.0%
Synonym(e):
Stannochlorid
About This Item
Lineare Formel:
SnCl2 · 2H2O
CAS-Nummer:
Molekulargewicht:
225.65
MDL-Nummer:
UNSPSC-Code:
12352302
PubChem Substanz-ID:
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Qualität
SAJ first grade
Assay
≥90.0%
Form
solid
Eignung der Reaktion
reagent type: catalyst
core: tin
Verfügbarkeit
available only in Japan
bp
652 °C (lit.)
mp (Schmelzpunkt)
37-38 °C (dec.) (lit.)
Lagertemp.
2-8°C
SMILES String
O.O.Cl[SnH2]Cl
InChI
1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2
InChIKey
FWPIDFUJEMBDLS-UHFFFAOYSA-L
Signalwort
Danger
Gefahreneinstufungen
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3
Zielorgane
Cardio-vascular system, Respiratory system
Lagerklassenschlüssel
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
David J Hallett et al.
Organic & biomolecular chemistry, 10(30), 6130-6158 (2012-04-20)
Reactions of the allyltin trichloride 45 generated from (4S)-4-benzyloxypent-2-enyl(tributyl)stannane 1 with imines prepared from glyoxylates proceed with useful levels of 1,5-stereocontrol in favour of (4E)-2,6-anti-2-(alkylamino)-6-benzyloxyhept-4-enoates 49. This stereoselectivity, controlled by the chirality of the stannane, dominates over any intrinsic stereochemical
Grégory Dupeyre et al.
Organic & biomolecular chemistry, 9(22), 7780-7790 (2011-10-07)
A six-step one-pot reaction was designed for synthesizing homodimeric 7-phenylindolo[3,2-a]carbazoles from 1H-indoles and β-nitrostyrenes, in the presence of SnCl(2)·2H(2)O. The reactions proceeded under very mild conditions and the desired heterocycles were obtained in moderate to good yields. An unprecedented mechanism
Rajendran Suresh et al.
The Journal of organic chemistry, 77(3), 1468-1476 (2012-01-11)
The synthesis of 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)quinolines by a SnCl(2)-catalyzed multicomponent reaction has been described. The reaction proceeds chemo- and regioselectively in an atom-economic way, generating a library of 24 quinoline derivatives.
Matthew Currie et al.
Inorganic chemistry, 52(4), 1710-1721 (2012-06-09)
The anionic speciation of chlorostannate(II) ionic liquids, prepared by mixing 1-alkyl-3-methylimidazolium chloride and tin(II) chloride in various molar ratios, χ(SnCl2), was investigated in both solid and liquid states. The room temperature ionic liquids were investigated by (119)Sn NMR spectroscopy, X-ray
Aili Sun et al.
Talanta, 88, 259-264 (2012-01-24)
The Zn-Sn nanoparticles/multiwall carbon nanotubes (Zn-SnNPs/MWNTs) nanocomposite film was prepared by electrodeposition of ZnCl(2) and SnCl(2) on MWNTs simultaneously in Ethaline ionic liquids. Then, based on immobilizing hemoglobin (Hb) within a novel Zn-SnNPs/MWNTs nanocomposite film (Hb/Zn-SnNPs/MWNTs), a novel hydrogen peroxide