Merck
P0SIC0NT1
Millipore
Milliflex® Positive Control Tool
To verify Milliflex® Rapid System Reagent Spray and Enumeration Functionality
About This Item
UNSPSC-Code:
41116500
NACRES:
NA.85
Materialien
aluminum
Verpackung
pkg of 1 kit
Hersteller/Markenname
Milliflex®
Methode(n)
Rapid Sterility Testing: suitable
Anwendung(en)
microbiology
pharmaceutical
sterility testing
Kompatibilität
for use with Milliflex® Rapid
Allgemeine Beschreibung
Milliflex® Positive Control Tool is designed to verify Milliflex® Rapid System reagent spray and microbial enumeration functionality.
Milliflex® Rapid systems are used to detect live microorganism in the tested sample. The positive control will help determine the efficiency of the reagent spray onto the sample and the corresponding microbial enumeration.
Anwendung
Milliflex® Positive Control Tool is used to verify Milliflex® Rapid System reagent spray and microbial enumeration functionality.
Rechtliche Hinweise
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Die Dokumentenbibliothek aufrufen
Artikel
Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.