Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NA.74
UNSPSC Code:
41103403
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
stainless steel holder
description
Compressed gas sampling
feature
autoclavable
manufacturer/tradename
MAS-100 Atmos®
application(s)
compressed gas monitoring
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
General description
MAS-100 Atmos® Plate Holder for MAS-100 Atmos® microbial compressed gas sampler is to place 90 mm agar plates appropriately under the perforated lid.
The lower part of the sampling head of the MAS-100 Atmos® is fixed, as same as with other mobile MAS-100® air samplers. Parts placed above are the instrument protection filter, filter holder, and plate holder. It is recommended not to remove these components before cleaning for routine applications. Disinfection can be done with clean tissue absorbed with 70% Ethanol or 70% IPA.
MAS-100 Atmos® Plate Holder is removable and autoclavable. In this case, the sampling head must be protected by the dust cover (separate accessory).
The lower part of the sampling head of the MAS-100 Atmos® is fixed, as same as with other mobile MAS-100® air samplers. Parts placed above are the instrument protection filter, filter holder, and plate holder. It is recommended not to remove these components before cleaning for routine applications. Disinfection can be done with clean tissue absorbed with 70% Ethanol or 70% IPA.
MAS-100 Atmos® Plate Holder is removable and autoclavable. In this case, the sampling head must be protected by the dust cover (separate accessory).
Application
The MAS-100 Atmos® Plate Holder is used to hold the 90 mm agar plates in the MAS-100 Atmos® Microbial Compressed Gas Sampler used for impaction.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Atmos is a registered trademark of MBV AG
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?