Merck
Z742438
MAGic Clamp magnetic clamp
holds 1 microplate
Größe auswählen
1 EA
€ 109,00
€ 109,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 109,00
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.73
€ 109,00
Check Cart for Availability
Leistungsmerkmale
holds 1 microplate
max. 4 xmicroplates
Verpackung
pkg of 1
Hersteller/Markenname
Benchmark Scientific H1000-MR-MP
Allgemeine Beschreibung
The convenient MAGic Clam platform for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
designed to be used with Benchmark Scientific H1010 shaking incubators and OrbiShaker XL orbital shakers
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)